Please login to your online account to display your discounted pricing

Magnesium foil, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Magnesium foil
CAS 7439-95-4
Physical Form ∽0.27g/25X25mm, Temper: As Rolled
Assay Percent Range 99.9% (metals basis)
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40603FF

  SDS
alfa aesar
40603FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40603FI

  SDS
alfa aesar
40603FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40603GH

  SDS
alfa aesar
40603GH
100 x 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form ∽0.27g/25X25mm, Temper: As Rolled
Quantity 25 x 25mm
EINECS Number 231-104-6
Chemical Name or Material Magnesium foil
CAS 7439-95-4
Assay Percent Range 99.9% (metals basis)
MDL Number MFCD00085308