Learn More
Magnesium ethoxide, Mg 21-22%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01426430
Description
Magnesium ethoxide is used as olefin polymerization catalyst carrier for polypropylene, polyethylene and precision ceramic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium ethoxide | |
270°C (decomposition) | |
1.01 g/mL | |
C4H10MgO2 | |
MFCD00009060 | |
UN3205 | |
Air and moisture sensitive | |
Insoluble in water. | |
CC[O-].CC[O-].[Mg+2] | |
114.427 | |
114.44 | |
Granules |
2414-98-4 | |
White | |
Odorless | |
Mg(OCH2CH3)2 | |
250 g | |
3678841 | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
magnesium;ethanolate | |
164963 | |
Mg 21 to 22% |
Chemical Identifiers
2414-98-4 | |
114.427 | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
164963 | |
CC[O-].CC[O-].[Mg+2] |
C4H10MgO2 | |
MFCD00009060 | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
magnesium;ethanolate |
Safety and Handling
GHS H Statement
H251-H319
Self-heating: may catch fire.
Causes serious eye irritation.
P210-P235+P410-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319-H500-X
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
EINECSNumber : 219-323-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only