missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium ethoxide, Mg 21-22%

Manufacturer:  Alfa Aesar™ 14264A1

 View more versions of this product

Catalog No. AA14264A1



Specifications

Specifications

Magnesium ethoxide
Odorless
White
1kg
C4H10MgO2
MFCD00009060
3678841
Mg 21 to 22%
CC[O-].CC[O-].[Mg+2]
114.427
114.44
1.01g/mL
Air & Moisture Sensitive
270°C (decomposition)
2414-98-4
Mg(OCH2CH3)2
UN3205
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
XDKQUSKHRIUJEO-UHFFFAOYSA-N
magnesium;ethanolate
164963
Granular

Chemical Identifiers

2414-98-4
114.427
XDKQUSKHRIUJEO-UHFFFAOYSA-N
164963
CC[O-].CC[O-].[Mg+2]
C4H10MgO2
MFCD00009060
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
magnesium;ethanolate
Safety and Handling

Safety and Handling

GHS H Statement
H251-H319
Self-heating: may catch fire.
Causes serious eye irritation.

GHS P Statement
P210-P280g-P305+P351+P338-P337+P313-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use ... for extinction.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.

TSCA : TSCA

Documents