missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium ethoxide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 397470050

 View more versions of this product

Catalog No. AC397470050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium ethoxide
270.0°C
Authentic
Glass bottle
(CH3CH2O)2Mg
5 g
Solubility in water: reacts
CC[O-].CC[O-].[Mg+2]
114.43
114.43
Powder
2414-98-4
White to Gray
98%
C4H10MgO2
MFCD00009060
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
XDKQUSKHRIUJEO-UHFFFAOYSA-N
magnesium;ethanolate
164963
98%

Chemical Identifiers

2414-98-4
114.43
XDKQUSKHRIUJEO-UHFFFAOYSA-N
164963
CC[O-].CC[O-].[Mg+2]
C4H10MgO2
MFCD00009060
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
magnesium;ethanolate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Self-heating; may catch fire.
Causes serious eye irritation.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
If

GHS Signal Word: Danger

EINECSNumber : 219-323-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only