Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium chloride, ultra dry, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Magnesium chloride
CAS 7786-30-3
Name Note Ultra dry
Linear Formula MgCl2
Formula Weight 95.22
MDL Number MFCD00011106
Merck Index 14,5662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3570909

  SDS
alfa aesar™
3570909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3570918

  SDS
alfa aesar™
3570918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3570930

  SDS
alfa aesar™
3570930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 95.22
Packaging Ampule
Mesh Size −10 Mesh
Merck Index 14,5662
Name Note Ultra dry
Physical Form Beads
Storage Note 1
  • Ampouled under argon
Chemical Name or Material Magnesium chloride
RTECS Number OM2800000
Assay Percent Notes (metals basis)
Density 2.32g/mL
Odor Odorless
Sensitivity Hygroscopic
Quantity 10g
Melting Point 714°C
TSCA TSCA
MDL Number MFCD00011106
Color White to yellow
CAS 7786-30-3
Linear Formula MgCl2
Assay 99.99%

  • Ampouled under argon

Safety and Handling

  • Ampouled under argon