Please login to your online account to display your discounted pricing

Magnesium chloride, ultra dry, 99.9% (metals basis), Alfa Aesar™

$42.50 - $373.68

Chemical Identifiers

Chemical Name or Material Magnesium chloride, ultra dry
CAS 7786-30-3
Physical Form Powder, Ampouled Under Argon
Assay Percent Range 99.9% (metals basis)
Molecular Formula Cl2Mg
Formula Weight 95.22
MDL Number MFCD00011106
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4284314

  SDS
alfa aesar
4284314
25g Each for $130.64

AA4284306

 
alfa aesar
4284306
5g Each for $42.50

AA4284322

  SDS
alfa aesar
4284322
100g Each for $373.68
Description & Specifications

Specifications

Formula Weight 95.22
Physical Form Powder, Ampouled Under Argon
Quantity 25g
EINECS Number 232-094-6
Density 2.32g/mL
Assay Percent Range 99.9% (metals basis)
Chemical Name or Material Magnesium chloride, ultra dry
Molecular Formula Cl2Mg
MDL Number MFCD00011106
CAS 7786-30-3