Please login to your online account to display your discounted pricing

Magnesium Chloride Reagent, For Total Cyanide, Certified, 51% (w/v), LabChem™

$88.63 - $401.31

Chemical Identifiers

Chemical Name or Material Magnesium Chloride Reagent
CAS 7732-18-5
CAS Max % 54.52
CAS 7791-18-6
CAS Max % 45.48
Name Note For Total Cyanide
Purity Grade Certified Grade
Linear Formula MgCl2·6H2O
Formula Weight 203.31
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC163901

 
lab chem
LC163901
500mL Each for $88.63

LC163902

 
lab chem
LC163902
1L Each for $128.97

LC163904

 
lab chem
LC163904
4L Each for $401.31
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 203.31
Chemical Name or Material Magnesium Chloride Reagent
Viscosity 2.7cSt
Decomposition Information Hydrogen chloride
Color Clear, colorless
Quantity 500mL
Name Note For Total Cyanide
Physical Form Liquid
Assay 51% (w/v)
Linear Formula MgCl2·6H2O
Density 1.19g/mL
CAS 7732-18-5
CAS Max % 54.52
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 7791-18-6
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 45.48
Solubility Soluble in water
Specific Gravity 1.177g/mL ±0.005
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.