missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Chloride Hexahydrate, (Crystalline),MP Biomedicals

$42.25 - $42.25

Chemical Identifiers

CAS 7791-18-6
Physical Form Crystalline
Molecular Formula MgCl2 · 6H2O
Formula Weight 203.3
Chemical Name or Material Magnesium Chloride Hexahydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19469880 Mp Biomedicals
19469880
100g Each for $42.25
Specifications Description & Specifications

Chemical Identifiers

7791-18-6
MgCl2 · 6H2O
Magnesium Chloride Hexahydrate
Crystalline
203.3

Specifications

7791-18-6
MgCl2 · 6H2O
100g
Crystalline
203.3
Magnesium Chloride Hexahydrate

Safety and Handling

Recommended Storage : Room Temperature