Please login to your online account to display your discounted pricing

Magnesium Chloride, Hexahydrate, ACS Grade, 99.0 to 102.0%, LabChem™

$119.80 - $119.80

Chemical Identifiers

Chemical Name or Material Magnesium Chloride
CAS 7791-18-6
CAS Max % 100
Name Note Hexahydrate
Purity Grade ACS Grade
Linear Formula MgCl2·6H2O
Formula Weight 203.31
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC163801

 
lab chem
LC163801
500g Each for $119.80
Description & Specifications

Specifications

Formula Weight 203.31
Chemical Name or Material Magnesium Chloride
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Assay 99.0 to 102.0%
Specific Gravity 1.569g/cm3
Storage Information Room Temperature
Linear Formula MgCl2·6H2O
Color White
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Density 1.569g/cm3
Name Note Hexahydrate
Physical Form Crystals
Solubility Soluble in water
CAS 7791-18-6
Decomposition Information Hydrogen chloride
Quantity 500g
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.