missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Chloride, Hexahydrate, ACS, 99.0 to 101.0%, EMD Millipore

$102.89 - $905.97

Chemical Identifiers

CAS 7791-18-6
Physical Form Solid
Assay Percent Range 99.0 to 101.0%
Molecular Formula Cl2Mg · 6H2O
Linear Formula MgCl2 · 6H2O
Formula Weight 23.3
Synonym Magnesium dichloride
Chemical Name or Material Magnesium Chloride, Hexahydrate, EMSURE™, For Analysis
Grade ACS/EP/ISO/Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1058330250 Emd Millipore Corporation
1058330250
250g Each for $102.89
M1058331000 Emd Millipore Corporation
1058331000
1kg Each for $293.17
M1058335000 Emd Millipore Corporation
1058335000
5kg Each for $905.97
Specifications Description & Specifications

Chemical Identifiers

7791-18-6
99.0 to 101.0%
MgCl2 · 6H2O
Magnesium dichloride
ACS/EP/ISO/Reagent Grade
Solid
Cl2Mg · 6H2O
23.3
Magnesium Chloride, Hexahydrate, EMSURE™, For Analysis

Specifications

23.3
White to gray white
1.57g/cm3 (20°C)
MgCl2 · 6H2O
Cl2Mg · 6H2O
Magnesium Chloride, Hexahydrate, EMSURE™, For Analysis
Solid
250g
Magnesium dichloride
99.0 to 101.0%
7791-18-6
ACS/EP/ISO/Reagent Grade

Safety and Handling

Recommended Storage : Store without restrictions