Learn More
Magnesium chloride hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197530250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium chloride hexahydrate | |
White | |
Crystalline Powder | |
98.5 to 101.5% (Complexometry) | |
MgCl2·6H2O | |
10,251; 15,196 | |
Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
203.30 | |
99% | |
Fibre drum |
117.0°C | |
7791-18-6 | |
25 kg | |
Cl2H12MgO6 | |
MFCD00149781 | |
15, 5726 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
magnesium(2+) hexahydrate dichloride | |
203.31 | |
Analytical |
Chemical Identifiers
7791-18-6 | |
203.30 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Cl2H12MgO6 | |
MFCD00149781 | |
magnesium(2+) hexahydrate dichloride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only