Please login to your online account to display your discounted pricing

Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

$224.58 - $224.58

Chemical Identifiers

Chemical Name or Material Magnesium Chloride Hexahydrate
CAS 7791-18-6
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range 99.0-101.0%
Molecular Formula MgCl2 · 6H2O
Formula Weight 203.3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19142101

 
mp biomedicals
19142101
1kg Each for $224.58
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 203.3
Quantity 1kg
Physical Form Crystalline
Chemical Name or Material Magnesium Chloride Hexahydrate
Assay Percent Range 99.0-101.0%
Molecular Formula MgCl2 · 6H2O
CAS 7791-18-6
Recommended Storage Room Temperature
Safety and Handling

Room Temperature