missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium Chloride, 0.0100 M, Ricca Chemical
Supplier: Ricca Chemical Company R4466000500A
Specifications
Magnesium Chloride Aqueous Solutions | |
Natural Poly Bottle | |
500 mL | |
0.01000 ± 0.00005 M at 20°C | |
Aqueous Solution |
0.01 M | |
7791-18-6,7732-18-5 | |
Colorless | |
Odorless | |
Solubility in Water: Infinite |