Please login to your online account to display your discounted pricing

Magnesium Chloride ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging Lined Fiber Dr

Manufacturer: avantor performance  595820

 View more versions of this product

Catalog No. MK5958A12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Lined Fiber Dr
Quantity 12kg
Chemical Name or Material Magnesium Chloride
Name Note 6-Hydrate
CAS 7791-18-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula MgCl2·6H2O
Formula Weight 203.3g/mol

Specifications

Chemical Name or Material Magnesium Chloride
Name Note 6-Hydrate
CAS 7791-18-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula MgCl2·6H2O
Formula Weight 203.3g/mol