Please login to your online account to display your discounted pricing

Magnesium Chloride ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  595802

 View more versions of this product

Catalog No. MK5958125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Magnesium Chloride
Name Note 6-Hydrate
CAS 7791-18-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula MgCl2·6H2O
Formula Weight 203.3g/mol

Specifications

Chemical Name or Material Magnesium Chloride
Name Note 6-Hydrate
CAS 7791-18-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula MgCl2·6H2O
Formula Weight 203.3g/mol