missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Chloride ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7791-18-6
Physical Form Crystalline
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
Chemical Name or Material Magnesium Chloride, 6-Hydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK5958212 Avantor Performance
595806
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK5958500 Avantor Performance
595804
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7791-18-6
MgCl2·6H2O
Magnesium Chloride, 6-Hydrate
Crystalline
203.3
ACS Grade

Specifications

7791-18-6
MgCl2·6H2O
Poly
Magnesium Chloride, 6-Hydrate
Crystalline
203.3
2.5kg
ACS Grade