Please login to your online account to display your discounted pricing

Magnesium Chloride, 6-Hydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$136.83 - $518.14

Chemical Identifiers

Chemical Name or Material Magnesium Chloride
CAS 7791-18-6
Name Note 6-Hydrate
Purity Grade Ultrapure Bioreagent
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003265

 
avantor performance
400301
Poly Bottle 500g Each for $136.83

02003266

 
avantor performance
400305
Poly Bottle 2.5kg Each for $518.14
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 203.3
Molecular Formula MgCl2·6H2O
CAS 7791-18-6
Name Note 6-Hydrate
Packaging Poly Bottle
Purity Grade Ultrapure Bioreagent
Specific Gravity 1L = 1.57kg
Quantity 500g
Chemical Name or Material Magnesium Chloride

    For Molecular Biology Applications