Please login to your online account to display your discounted pricing

Magnesium Chloride, 6-Hydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  400305

 View more versions of this product

Catalog No. 02003266

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Magnesium Chloride
Name Note 6-Hydrate
CAS 7791-18-6
Purity Grade Ultrapure Bioreagent
Physical Form Crystals
Molecular Formula MgCl2·6H2O
Formula Weight 203.3
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Quantity 2.5kg

Specifications

Chemical Name or Material Magnesium Chloride
CAS 7791-18-6
Name Note 6-Hydrate
Purity Grade Ultrapure Bioreagent
Molecular Formula MgCl2·6H2O
Formula Weight 203.3

    For Molecular Biology Applications