Please login to your online account to display your discounted pricing

Magnesium Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  244405

 View more versions of this product

Catalog No. 02003263

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 203.3
Physical Form Crystals
CAS 7791-18-6
Specific Gravity 1L = 1.57kg
Molecular Formula MgCl2·6H2O
Name Note 6-Hydrate
Quantity 2.5kg
Packaging Poly Bottle
Chemical Name or Material Magnesium Chloride

Specifications

Chemical Name or Material Magnesium Chloride
CAS 7791-18-6
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula MgCl2·6H2O
Formula Weight 203.3