Please login to your online account to display your discounted pricing

Magnesium chloride, -10 Mesh Beads, ultra dry, 99.9% (metals basis), Alfa Aesar

Chemical Identifiers

Chemical Name or Material Magnesium chloride, ultra dry
CAS 7786-30-3
Molecular Formula Cl2Mg
Formula Weight 95.22
MDL Number MFCD00011106
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3570718

 
alfa aesar™
3570718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder, Ampouled Under Argon
Formula Weight 95.22
Quantity 50g
Chemical Name or Material Magnesium chloride, ultra dry
EINECS Number 232-094-6
Assay Percent Range 99.9% (metals basis)
Molecular Formula Cl2Mg
MDL Number MFCD00011106
CAS 7786-30-3
Density 2.32g/mL