Please login to your online account to display your discounted pricing

Magnesium Carbonate (Powder/USP/FCC), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLM27500

 View more versions of this product

Catalog No. M27-500

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 84.31
Melting Point 350°C
Quantity 500g
Density 3.04g/mL
Physical Form Solid
Assay 40.0 to 43.5% (USP)
Molecular Formula CMgO3
Viscosity [NULL]
Decomposition Information 350°C
Color White
Heavy Metals (as Pb) 0.003% max. (USP)
Identification Pass Test (FCC)
Insoluble in Acid 0.05% max. (FCC)
Residual Solvents Meets Requirements (USP)
Arsenic (As) 4ppm max. (USP)
Calcium (Ca) 0.45% max. (USP)
Calcium Oxide 0.6% max. (FCC)
Lead (Pb) 10mg/kg max. (FCC)
Packaging Poly Bottle
MDL Number MFCD00064632
CAS 546-93-0
ChemAlert Storage Symbol Gray
Chemical Name or Material Magnesium Carbonate, Hydrated
Iron (Fe) 0.02% max. (USP)
Soluble Salts 1% max. (FCC)
E. coli Negative (USP)

Specifications

Chemical Name or Material Magnesium Carbonate, Hydrated
CAS 546-93-0
Purity Grade FCC/USP
Physical Form Solid
Assay 40.0 to 43.5% (USP)
Molecular Formula CMgO3
Formula Weight 84.31
MDL Number MFCD00064632
Accessories
 

SDS & Safety Documents