Please login to your online account to display your discounted pricing

Magnesium Carbonate (Powder/USP/FCC), Fisher Chemical

$304.43 - $652.36

Chemical Identifiers

Chemical Name or Material Magnesium Carbonate, Hydrated
CAS 546-93-0
Purity Grade FCC/USP
Physical Form Solid
Assay 40.0 to 43.5% (USP)
Molecular Formula CMgO3
Formula Weight 84.31
MDL Number MFCD00064632
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M27-212

  SDS
fisher scientific company
FLM27212
Poly Pail 2.5kg Each for $652.36

M27-500

  SDS
fisher scientific company
FLM27500
Poly Bottle 500g Each for $304.43
Case of 4 Each for $1,217.72
Description & Specifications

Specifications

Purity Grade FCC/USP
Formula Weight 84.31
Melting Point 350°C
Quantity 2.5kg
Assay 40.0 to 43.5% (USP)
Physical Form Solid
Color White
Molecular Formula CMgO3
Decomposition Information 350°C
Residual Solvents Meets Requirements (USP)
Insoluble in Acid 0.05% max. (FCC)
Arsenic (As) 4ppm max. (USP)
Heavy Metals (as Pb) 0.003% max. (USP)
Soluble Salts 1% max. (FCC)
Chemical Name or Material Magnesium Carbonate, Hydrated
Identification Pass Test (FCC)
ChemAlert Storage Symbol Gray
CAS 546-93-0
MDL Number MFCD00064632
Iron (Fe) 0.02% max. (USP)
Calcium Oxide 0.6% max. (FCC)
Lead (Pb) 10mg/kg max. (FCC)
E. coli Negative (USP)
Calcium (Ca) 0.45% max. (USP)
Packaging Poly Pail
Safety and Handling