missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium carbonate hydroxide hydrate, Puratronic™, 99.996% (metals basis)

Manufacturer:  Alfa Aesar™ 1079622

 View more versions of this product

Catalog No. AA1079622



Specifications

Specifications

Magnesium carbonate hydroxide hydrate
(metals basis)
14,5660
142.63 (Anhydrous)
99.996%
White
39409-82-0
MFCD00149783
Magnesium carbonate, basic; Magnesium hydroxide carbonate
Powder
Puratronic™
100g

Chemical Identifiers

39409-82-0
MFCD00149783
Magnesium carbonate, basic; Magnesium hydroxide carbonate
Safety and Handling

Safety and Handling

RTECSNumber : FG1797000

Documents