missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211070025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Magnesium carbonate basic, light | |
2.5kg | |
Mg(OH)2·5H2O | |
15, 5724 | |
121.41 | |
40.0 to 45.0% (MgO) | |
Plastic drum | |
2.1600g/mL |
39409-82-0 | |
1/4H2O2Mg·5/4H2O | |
MFCD07371358 | |
Solubility in water: practically insoluble | |
121.41 | |
Analysis | |
2.16 |
RUO â Research Use Only