Please login to your online account to display your discounted pricing

Magnesium bromide hexahydrate, 99+%, for analysis, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC202510050

 View more versions of this product

Catalog No. AC202510050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99%
Physical Form Crystalline Powder
Formula Weight 292.21
Quantity 5g
Color White
Merck Index 14, 5659
Purity Grade For analysis
pH 6.5 to 8 (5% soln. at 20°C)
Fieser 13,259; 15,194; 16,199
Name Note p.a.
Linear Formula MgBr2·6H2O
Sulfate 200ppm max.
CAS 13446-53-2
Trace Analysis 50ppm max.
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Cadmium (Cd) 50ppm max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Magnesium bromide hexahydrate
Assay Percent Range 99+%
Molecular Formula Br2Mg·6H2O
MDL Number MFCD00149780
Iron (Fe) 50ppm max.
Chlorine (Cl) 500ppm max.
Copper (Cu) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Glass bottle

Specifications

Chemical Name or Material Magnesium bromide hexahydrate
Name Note p.a.
CAS 13446-53-2
Purity Grade For analysis
Assay ≥99%
Assay Percent Range 99+%
Molecular Formula Br2Mg·6H2O
Linear Formula MgBr2·6H2O
Formula Weight 292.21
MDL Number MFCD00149780
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents