Learn More
Magnesium bromide hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202512500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium bromide hexahydrate | |
p.a. | |
White | |
Crystalline Powder | |
99% min. (Complexometry) | |
MgBr2·6H2O | |
13,259; 15,194; 16,199 | |
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent | |
LGLXXNHIGIJYQQ-UHFFFAOYSA-L | |
dibromomagnesium hexahydrate | |
17749097 | |
≥99% | |
Glass bottle |
165.0°C | |
13446-53-2 | |
6.5 to 8 (5% soln. at 20°C) | |
250 g | |
Br2H12MgO6 | |
MFCD00149780 | |
14, 5659 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
O.O.O.O.O.O.Br[Mg]Br | |
292.20 | |
292.21 | |
Analytical |
Chemical Identifiers
13446-53-2 | |
292.20 | |
LGLXXNHIGIJYQQ-UHFFFAOYSA-L | |
17749097 | |
O.O.O.O.O.O.Br[Mg]Br |
Br2H12MgO6 | |
MFCD00149780 | |
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent | |
dibromomagnesium hexahydrate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
RUO – Research Use Only