missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium bromide hexahydrate, 98+%

Manufacturer:  Alfa Aesar™ 4612022

 View more versions of this product

Catalog No. AA4612022



Specifications

Specifications

Magnesium bromide hexahydrate
Br2H12MgO6
MFCD00149780
Soluble in water, somewhat soluble in alcohol
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
292.203
292.22 (184.13 Anhydrous)
≥98%
Odorless
Colorless
100g
13446-53-2
MgBr2·6H2O
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
magnesium;dibromide;hexahydrate
17749097
Crystalline
2.00g/mL
Hygroscopic
172°C

Chemical Identifiers

13446-53-2
292.203
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
17749097
O.O.O.O.O.O.[Mg+2].[Br-].[Br-]
Br2H12MgO6
MFCD00149780
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide
magnesium;dibromide;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents