Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium bromide diethyl etherate, 98%

$62.68 - $62.68

Chemical Identifiers

Chemical Name or Material Magnesium bromide diethyl etherate
CAS 29858-07-9
Molecular Formula C4H10O·Br2Mg
Linear Formula (CH3CH2)2O·MgBr2
Formula Weight 258.24
MDL Number MFCD00064500
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1548114

  SDS
alfa aesar™
L1548114
25g Each for $62.68

AAL1548122

  SDS
alfa aesar™
L1548122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.24
Chemical Name or Material Magnesium bromide diethyl etherate
CAS 29858-07-9
Quantity 25g
Flash Point 35°C (95°F)
UN Number UN1325
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Assay 98%
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Molecular Formula C4H10O·Br2Mg
MDL Number MFCD00064500
Health Hazard 1 Danger
Linear Formula (CH3CH2)2O·MgBr2

Applications
The magnesium bromide diethyl etherate (MgBr2· OEt2)-methyl sulfide (Me2S) system is useful for the mild and chemo-selective deprotection of p-methoxybenzyl (PMB) ether. Lewis acid for catalysis of Diels-Alder reactions of homochiral 2-sulfinylmaleates with cyclopentadiene. Magnesium bromide diethyl etherate is used in the synthesis of silicon-containing azole derivatives.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.