missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 4666706

 View more versions of this product

Catalog No. AA4666706



Specifications

Specifications

Magnesium bis (trifluoromethylsulfonyl)imide
White to Yellow
5g
C4F12MgN2O8S4
MFCD01321278
Magnesium triflimide; Magnesium bis(trifluoromethanesulfonimide)
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]
584.576
584.58
Moisture Sensitive
>200°C
133395-16-1
[(CF3SO2)2N]2Mg
UN1759
DMFBPGIDUUNBRU-UHFFFAOYSA-N
magnesium;bis(trifluoromethylsulfonyl)azanide
11851070
Solid

Chemical Identifiers

133395-16-1
584.576
DMFBPGIDUUNBRU-UHFFFAOYSA-N
11851070
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]
C4F12MgN2O8S4
MFCD01321278
Magnesium triflimide; Magnesium bis(trifluoromethanesulfonimide)
magnesium;bis(trifluoromethylsulfonyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

SDS Documents