Please login to your online account to display your discounted pricing

Magnesium bis(diisopropylamide), 0.7M solution in THF, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC389468000

 View more versions of this product

Catalog No. AC389468000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 224.68
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
Concentration 0.6 to 0.8M
Density 0.8800g/mL
Chemical Name or Material Magnesium bis(diisopropylamide)
Appearance of Solution Yellowish-brown solution
Name Note 0.7M solution in THF
Molecular Formula C12H28MgN2
MDL Number MFCD03093019
CAS 23293-23-4
Health Hazard 1 Danger
CAS 109-99-9
CAS na
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
CAS 513-35-9

Specifications

Chemical Name or Material Magnesium bis(diisopropylamide)
Name Note 0.7M solution in THF
CAS 23293-23-4
CAS 109-99-9
CAS 513-35-9
CAS na
Molecular Formula C12H28MgN2
Formula Weight 224.68
MDL Number MFCD03093019
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.

SDS & Safety Documents