Please login to your online account to display your discounted pricing

Magnesium bis(diisopropylamide), 0.7M solution in THF, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Magnesium bis(diisopropylamide)
Name Note 0.7M solution in THF
CAS 23293-23-4
CAS 109-99-9
CAS 513-35-9
CAS na
Molecular Formula C12H28MgN2
Formula Weight 224.68
MDL Number MFCD03093019
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389461000

  SDS
thermo fisher scientific
AC389461000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC389468000

  SDS
thermo fisher scientific
AC389468000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.68
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.8800g/mL
Concentration 0.6 to 0.8M
Appearance of Solution Yellowish-brown solution
CAS 513-35-9
CAS 109-99-9
CAS na
CAS 23293-23-4
Chemical Name or Material Magnesium bis(diisopropylamide)
MDL Number MFCD03093019
Name Note 0.7M solution in THF
Molecular Formula C12H28MgN2
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Safety and Handling

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.