Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium aluminum zirconium oxide, 99% (metals basis excluding Ca) Ca 0.1-1%

Chemical Identifiers

Chemical Name or Material Magnesium aluminum zirconium oxide
CAS 53169-11-2
Formula Weight 265.46g/mol
MDL Number MFCD00171346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3636814

 
alfa aesar™
36368-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3636822

 
alfa aesar™
36368-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Particle Size 3 to 12mm
Quantity 25g
Chemical Name or Material Magnesium aluminum zirconium oxide
CAS 53169-11-2
Physical Form Sintered Lump
Assay 99%
Assay Percent Notes (metals basis excluding Ca) Ca 0.1 to 1%
Formula Weight 265.46g/mol
MDL Number MFCD00171346