Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long

Chemical Identifiers

Chemical Name or Material Magnesium Aluminum Zinc wire
Name Note 3.18mm (0.125 in.) dia., 90cm (35 in.) long
MDL Number MFCD01074901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14092RN

  SDS
alfa aesar™
14092RN
5 x 90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14092RO

  SDS
alfa aesar™
14092RO
25x90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14092RP

  SDS
alfa aesar™
14092RP
100x90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Magnesium Aluminum Zinc wire
TSCA TSCA
Name Note 3.18mm (0.125 in.) dia., 90cm (35 in.) long
Physical Form Wire
MDL Number MFCD01074901
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Mg:Al:Zn; 93:6:1 wt%
Weight ≈12.44g/90cm
Quantity 5 x 90cm