Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium Aluminum Zinc wire, 1.59mm (0.063 in.) dia., 90cm (35 in.) long

Chemical Identifiers

Chemical Name or Material Magnesium Aluminum Zinc wire
Name Note 1.59mm (0.063 in.) dia., 90cm (35 in.) long
MDL Number MFCD01074901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14069RN

  SDS
alfa aesar™
14069RN
5 x 90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14069RO

  SDS
alfa aesar™
14069RO
25x90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14069RP

  SDS
alfa aesar™
14069RP
100x90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Mg:Al:Zn; 93:6:1 wt%
Physical Form Wire
Chemical Name or Material Magnesium Aluminum Zinc wire
Name Note 1.59mm (0.063 in.) dia., 90cm (35 in.) long
TSCA TSCA
Odor Odorless
MDL Number MFCD01074901
Weight ≈3.1g/90cm
Quantity 5 x 90cm