Alfa Aesar™ Magnesium Aluminum Zinc wire, 1.59mm (0.063 in.) dia., 90cm (35 in.) long
Manufacturer:  Alfa Aesar™ 14069RP
 View more versions of this product
Catalog No. AA14069RP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Mg:Al:Zn; 93:6:1 wt%
≈3.1g/90cm
1.59mm (0.063 in.) dia., 90cm (35 in.) long
Odorless
Magnesium Aluminum Zinc wire
100 x 90cm
MFCD01074901
Wire
Documents