Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium Aluminum Zinc plate, alloy AZ31B, 6.35mm (0.25 in.) thick

Chemical Identifiers

Chemical Name or Material Magnesium Aluminum Zinc plate, Alloy AZ31B
Name Note 6.35mm (0.25 in.) thick
MDL Number MFCD01074901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14066RF

  SDS
alfa aesar™
14066RF
30x30cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Magnesium Aluminum Zinc plate, Alloy AZ31B
TSCA TSCA
Name Note 6.35mm (0.25 in.) thick
Physical Form Plate
MDL Number MFCD01074901
Odor Odorless
Avg. Mol. Wt. or Mol. Wt. Range Mg:Al:Zn; 96:3:1 wt%
Weight ≈994g/30x30cm
Quantity 30x30cm