missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium aluminum oxide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 2295022

 View more versions of this product

Catalog No. AA2295022



Specifications

Specifications

Magnesium aluminum oxide
99%
Al2MgO4
Magnesium aluminate
142.26
Powder
Odorless
12068-51-8
(metals basis)
MFCD00049475
UAMZXLIURMNTHD-UHFFFAOYSA-N
142.26
−325 Mesh
100g

Chemical Identifiers

12068-51-8
142.26
UAMZXLIURMNTHD-UHFFFAOYSA-N
Al2MgO4
MFCD00049475
Magnesium aluminate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents