Learn More
Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03635822
Description
Used as a source of magnesium in biological reactions and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium acetate tetrahydrate | |
16674-78-5 | |
Crystals | |
98.0-102.0% | |
MFCD00149214 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
magnesium(2+) diacetate tetrahydrate | |
134717 | |
ACS | |
Hygroscopic | |
1.454 g/cm3 at 20°C |
80°C | |
White | |
100 g | |
C4H14MgO8 | |
3730605 | |
Soluble in water. | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
214.45 | |
214.45 (142.39 Anhydrous) | |
Odorless | |
White crystals |
Chemical Identifiers
16674-78-5 | |
214.45 | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
134717 | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
C4H14MgO8 | |
MFCD00149214 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
magnesium(2+) diacetate tetrahydrate |
Safety and Handling
H500
EINECSNumber : 205-554-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only