Please login to your online account to display your discounted pricing

Magnesium acetate tetrahydrate, 99.997% (metals basis), Alfa Aesar™, Puratronic™

Chemical Identifiers

Chemical Name or Material Magnesium acetate tetrahydrate
CAS 16674-78-5
Physical Form Crystals
Assay Percent Range 99.997% (Metal basis)
Molecular Formula Mg(CH3COO)2·4H2O
Formula Weight 214.45 (142.39anhydrous)
MDL Number MFCD00149214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1079514

  SDS
alfa aesar
1079514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1079522

  SDS
alfa aesar
1079522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Magnesium acetate tetrahydrate
CAS 16674-78-5
EINECS Number 205-554-9
Physical Form Crystals
Assay Percent Range 99.997% (Metal basis)
Molecular Formula Mg(CH3COO)2·4H2O
Appearance White crystals
Formula Weight 214.45 (142.39 Anhydrous)
MDL Number MFCD00149214
Density 1.454g/cm3 at 20°C
Odor Odorless
Color White
Quantity 25g