missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium acetate tetrahydrate, 99.997% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1079514

 View more versions of this product

Catalog No. AA1079514



Specifications

Specifications

Magnesium acetate tetrahydrate
99.997% (Metal basis)
MFCD00149214
acetic acid, magnesium salt, tetrahydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
214.453
214.45 (142.39 Anhydrous)
1.454g/cm3 at 20°C
White
16674-78-5
C4H14MgO8
White crystals
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium;diacetate;tetrahydrate
134717
Crystals
Odorless
25g

Chemical Identifiers

16674-78-5
214.453
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
C4H14MgO8
MFCD00149214
acetic acid, magnesium salt, tetrahydrate
magnesium;diacetate;tetrahydrate
Safety and Handling

Safety and Handling

EINECSNumber : 205-554-9

Documents