missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium acetate tetrahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
$169.10 - $452.98
Chemical Identifiers
CAS | 16674-78-5 |
---|---|
Molecular Formula | C4H14MgO8 |
Molecular Weight (g/mol) | 214.45 |
MDL Number | MFCD00149214 |
InChI Key | XKPKPGCRSHFTKM-UHFFFAOYSA-L |
Synonym | magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p |
PubChem CID | 134717 |
SMILES | O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC270500050
|
Thermo Scientific Chemicals
270500050 |
5 g | Glass Bottle |
Each for $169.10
|
|
||||
AC270500010
|
Thermo Scientific Chemicals
270500010 |
1 kg | Plastic Bottle |
Each for $452.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
16674-78-5 | |
214.45 | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
134717 |
C4H14MgO8 | |
MFCD00149214 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
Specifications
80.0°C | |
White | |
Crystalline Powder | |
99.5% min. (Complexometry) | |
(CH3CO2)2Mg·4H2O | |
15,196 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
magnesium(2+) diacetate tetrahydrate | |
134717 | |
99.5% | |
Authentic | |
Magnesium acetate tetrahydrate, p.a. |
16674-78-5 | |
8 to 9 (5% soln. at 20°C) | |
5 g | |
C4H14MgO8 | |
MFCD00149214 | |
15,5718 | |
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
214.45 | |
214.46 | |
Analytical | |
Glass Bottle |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only