missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium acetate tetrahydrate, 98%

Manufacturer:  Alfa Aesar™ 12225A1

 View more versions of this product

Catalog No. AA12225A1



Specifications

Specifications

Magnesium acetate tetrahydrate
C4H14MgO8
3730605
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium;diacetate;tetrahydrate
134717
Crystalline
1.454g/mL
Hygroscopic
1kg
16674-78-5
MFCD00149214
14,5654
Freely soluble in water, alcohol
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
214.453
214.45 (142.39 Anhydrous)
98%
Odorless
80°C

Chemical Identifiers

16674-78-5
214.453
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
magnesium;diacetate;tetrahydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents