missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline, ACROS Organics™

Manufacturer:  Acros Organics AC211570051

 View more versions of this product

Catalog No. AC211570051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Magnesium acetate tetrahydrate
16674-78-5
C4H14MgO8
MFCD00149214
15, 5718
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium;diacetate;tetrahydrate
134717
Crystalline Powder
Extra Pure
20ppm max.
50ppm max.
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol
White
5kg
Extra Pure
97.5%
(CH3CO2)2Mg·4H2O
15,196
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
214.453
214.46
97.5%
100ppm max.
Authentic
Plastic drum
500ppm max.
80.0°C

Chemical Identifiers

16674-78-5
214.453
XKPKPGCRSHFTKM-UHFFFAOYSA-L
134717
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mg+2]
C4H14MgO8
MFCD00149214
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
magnesium;diacetate;tetrahydrate
SDS Documents