Please login to your online account to display your discounted pricing

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  211572500

 View more versions of this product

Catalog No. AC211572500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 214.46
Physical Form Crystalline Powder
Assay ≥97%
Purity Grade Extra Pure
Quantity 250g
Color White
Merck Index 15, 5718
Fieser 15,196
Iron (Fe) 50ppm max.
Linear Formula (CH3CO2)2Mg·4H2O
Heavy Metals (as Pb) 20ppm max.
Molecular Formula C4H6O4Mg·4H2O
Name Note Extra Pure
Chemical Name or Material Magnesium acetate tetrahydrate
Assay Percent Range 97.5%
CAS 16674-78-5
MDL Number MFCD00149214
Synonym Acetic acid, magnesium salt tetrahydrate
Packaging Plastic bottle
Sulfate 500ppm max.
Chlorine (Cl) 100ppm max.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Magnesium acetate tetrahydrate
Name Note Extra Pure
CAS 16674-78-5
Purity Grade Extra Pure
Physical Form Crystalline Powder
Assay ≥97%
Assay Percent Range 97.5%
Molecular Formula C4H6O4Mg·4H2O
Linear Formula (CH3CO2)2Mg·4H2O
Formula Weight 214.46
MDL Number MFCD00149214
Synonym Acetic acid, magnesium salt tetrahydrate