missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Acetate GenAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 16674-78-5
Molecular Formula (CH3COO)2Mg·4H2O
Formula Weight 214.45
Chemical Name or Material Magnesium Acetate, 4-Hydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7799500 Avantor Performance
779904
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK7799212 Avantor Performance
779906
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16674-78-5
(CH3COO)2Mg·4H2O
214.45
Magnesium Acetate, 4-Hydrate

Specifications

214.45
POLYSTORMOR
500g
(CH3COO)2Mg·4H2O
16674-78-5
Magnesium Acetate, 4-Hydrate