Please login to your online account to display your discounted pricing

Magnesium Acetate GenAR, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  779904

 View more versions of this product

Catalog No. MK7799500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 214.45
Molecular Formula (CH3COO)2Mg·4H2O
Name Note 4-Hydrate
Chemical Name or Material Magnesium Acetate
Packaging POLYSTORMOR
CAS 16674-78-5
Quantity 500g

Specifications

Chemical Name or Material Magnesium Acetate
Name Note 4-Hydrate
CAS 16674-78-5
Molecular Formula (CH3COO)2Mg·4H2O
Formula Weight 214.45