missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium, AAS standard solution, Specpure™, Mg 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 88077AE

 View more versions of this product

Catalog No. AA88077AE



Specifications

Specifications

Magnesium
Mg
Nitric acid: 5%; Magnesium: 0.1%; Water: 94.9%
[Mg]
24.305
CHEBI:25107
17mm Hg at 20°C
100mL
7439-95-4,7697-37-2,7732-18-5
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
5462224
Liquid
Colorless

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
CHEBI:25107
[Mg]
Mg
MFCD00085308
5462224
magnesium
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents