Please login to your online account to display your discounted pricing

Magnesium, AAS standard solution, Specpure™, Mg 1000μg/mL, Alfa Aesar™

$95.43 - $95.43

Chemical Identifiers

Chemical Name or Material Magnesium
CAS 7697-37-2
CAS 7439-95-4
CAS 7732-18-5
Physical Form Liquid
Molecular Formula Mg in 5% HNO3
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88077AE

 
alfa aesar
88077AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88077AP

 
alfa aesar
88077AP
500mL Each for $95.43
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Magnesium
EINECS Number 231-714-2
Molecular Formula Mg in 5% HNO3
MDL Number MFCD00085308
CAS 7697-37-2
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Magnesium: 0.1%; Water: 94.9%
Health Hazard 1 H314: Causes severe skin burns and eye damage.
CAS 7439-95-4
Vapor Pressure 17mm Hg at 20°C
CAS 7732-18-5
Safety and Handling

H314: Causes severe skin burns and eye damage.