missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium AA Standard, 1000 ppm Mg in 3% HNO3, Ricca Chemical
$110.94 - $298.09
Chemical Identifiers
CAS | 7439-95-4 |
---|---|
Molecular Formula | Mg |
Molecular Weight (g/mol) | 24.30 |
MDL Number | MFCD00085308 |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
PubChem CID | 5462224 |
ChEBI | CHEBI:25107 |
IUPAC Name | magnesium |
SMILES | [Mg] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AMG1KN-100
|
Ricca Chemical Company
AMG1KN100 |
100 mL | Natural LDPE Bottle |
Each for $110.94
|
|
||||
AMG1KN-500
|
Ricca Chemical Company
AMG1KN500 |
500 mL | Natural Poly Bottle |
Each for $298.09
|
|
||||
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
CHEBI:25107 | |
[Mg] |
Mg | |
MFCD00085308 | |
5462224 | |
magnesium |
Specifications
7439-95-4,7697-37-2,7732-18-5 | |
100 mL | |
97.0,5.09,0.1 | |
1,000ppm Mg | |
MFCD00085308 | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
Liquid |
Natural LDPE Bottle | |
93.0,4.89,0.09 | |
Colorless to Yellow | |
Mg | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
1 mL = 1 mg Mg | |
Magnesium AA Standard |