Please login to your online account to display your discounted pricing

American Educational Products Magnesium

$17.90 - $17.90

Chemical Identifiers

Chemical Name or Material Magnesium
CAS 7439-95-4
Linear Formula Mg
Formula Weight 24.31g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S75532

 
american educational products
7-807
Metal Ribbon Each for $17.90
Description & Specifications

Specifications

Packaging Metal Ribbon
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Color Silver-gray
Chemical Name or Material Magnesium
CAS 7439-95-4
Linear Formula Mg
Formula Weight 24.31g/mol
Shelf Life Indefinite
ChemAlert Storage Symbol Red
DOT Information DOT Class 4.1, : Flammable Solid
Safety and Handling
 

Indefinite